Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oligomycin, MP Biomedicals™

Catalog No. MP215178625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215178625 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215178625 Supplier MP Biomedicals, Inc Supplier No. 0215178625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits mitochondrial ATPase and phosphoryl group transfer. Oligomycin, MP Biomedicals is a natural antibiotic isolated from Streptomyces diastatochromogenes. It is a mixture of oligomycins A, B, and C.

Oligomycin is used as antibiotic which inhibits respiration in mitochondria. It inhibits ATP synthase by blocking its proton channel (F0 subunit), which is necessary for oxidative phosphorylation of ADP to ATP (energy production).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1404-19-9
Molecular Formula C45H74O11
Molecular Weight (g/mol) 791.08
MDL Number MFCD00065705,MFCD01779388
InChI Key MNULEGDCPYONBU-ZUSSGZTJNA-N
Synonym Olivomycin, Aburamycin
PubChem CID 78358496
IUPAC Name (4E,18E,20Z)-22-ethyl-7,11,14,15-tetrahydroxy-6'-(2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione
SMILES CCC1CCC2OC3(CCC(C)C(CC(C)O)O3)C(C)C(OC(=O)\C=C\C(C)C(O)C(C)C(=O)C(C)C(O)C(C)C(=O)C(C)(O)C(O)C(C)C\C=C\C=C/1)C2C

Specifications

Type Oligomycin
Physical Form Powder
Melting Point 150°C (literature)
Source Streptomyces diastatochromogenes
Quantity 25 mg
Solubility Information Soluble in ether, benzene, absolute ethanol, glacial acetic acid and acetone.
Formula Weight ∼789.3
Grade Not Available
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.