accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1404-19-9

IUPAC Name: (4E,18E,20Z)-22-ethyl-7,11,14,15-tetrahydroxy-6'-(2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione
Synonyms: oligomycin
Molecular Weight (g/mol): 791.08
Molecular Formula: C45H74O11
InChi Key: MNULEGDCPYONBU-ZUSSGZTJNA-N
SMILES: CCC1CCC2OC3(CCC(C)C(CC(C)O)O3)C(C)C(OC(=O)\C=C\C(C)C(O)C(C)C(=O)C(C)C(O)C(C)C(=O)C(C)(O)C(O)C(C)C\C=C\C=C/1)C2C
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (4)
Type 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (4)