Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLLAS Tag Monoclonal Antibody (L2)
GREENER_CHOICE

Catalog No. PIMA516125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516125 Supplier Invitrogen™ Supplier No. MA516125
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

OLLAS Tag Monoclonal Antibody for ICC/IF, IHC (F), IHC (P), IP, Western Blot

The OLLAS (E.coli OmpF Linker and mouse Langerin fusion Sequence) epitope tag is a newly defined, highly sensitive tag. It is a superior tag for immunoprecipitation, fluorescent immunohistochemistry and flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLLAS Tag
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone L2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Host Species Rat
Immunogen OLLAS tagged extracellular domain of mouse Langerin.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.