Learn More
TARGETMOL CHEMICALS INC OLMUTINIB 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6918
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olmutinib (HM61713 BI 1482694) is an orally available small molecule a mutant-selective inhibitor of epidermal growth factor receptor (EGFR) with potential antineoplastic activity. Olmutinib binds to and inhibits mutant forms of EGFR thereby leading to cell death of EGFR-expressing tumor cells. As this agent is selective towards mutant forms of EGFR its toxicity profile may be reduced as compared to non-selective EGFR inhibitors which also inhibit the EGFR wild-type form. purity: 99%
Catalog No. 50-257-6415
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.