Learn More
Ansell Olympic Optical Code 4 Eyewear
Catalog No. 191301612 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Black
Platinum
Lens Tint:
Mirror
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-130-1612 Black Mirror
19-130-1610 Platinum Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-1612 Supplier Ansell Supplier No. 19834
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Olympic Optical Code 4 Eyewear provide all-day comfort and protection, for both work and play
  • Stylish, minimal black frames hold lenses with a 7 base curve
  • Ribbed nose cushion helps keep the glasses from slipping
  • Contoured temple tips give the wearer a precise fit
  • Hinge stop helps prevent lens damage, so the protective glasses are really built to last
  • Provide 99.9% UVA / UVB / UVC protection and meet ANSI Z87.1+ standards for high impact
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meets ANSI Z87.1+ standards
Lens Tint Mirror
Product Type Safety Glasses
Frame Color Black
No. per Pack 12/Pk.
Lens Material Polycarbonate