Learn More
Ansell Olympic Optical Smith and Wesson 38 Special Eyewears
Catalog No. 191304577 Shop All Ansell Products
Change view
Click to view available options
:
Olympic Optical Smith and Wesson 38 Special Eyewears
Catalog No.
19-130-4577 Olympic Optical Smith and Wesson 38 Special Eyewears
1 options
This item is not returnable. View return policy
Catalog No. 19-130-4577 Supplier Ansell Supplier No. 19857
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Olympic Optical Smith and Wesson 38 Special Eyewears are compactly designed and have a variety of lens and frame options
  • Ergonomically designed temples with rubber insert temple tips for a soft, snug and secure fit
  • All-day comfort
  • 8 base curve lenses are hard coated for scratch resistance and longer life
  • 99.9% UV (UVA / UVB / UVC) protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Meet the high impact level for the ANSI Z87.1+ standards for personal eye protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1+
Lens Tint Smoke
Product Type Safety Glasses
Temple Style Sure-grip rubber