Learn More
MilliporeSigma™ Supelco™ Ombifit™ Small Bore Connector Three-way valve connector
Catalog No. 111010129 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Includes PTFE cones
Catalog No.
11-101-0129 Includes PTFE cones
1 options
Catalog No. 11-101-0129 Supplier MilliporeSigma™ Supelco™ Supplier No. 56142
Add to Cart
Add to Cart
Omnifit™ Small Bore Connectors offer the unique advantage of producing a very low dead volume connection with tubing of various diameters, through an O-ring compression system, a cone over tubing system or a void-filling cone with a standard gripper.
TRUSTED_SUSTAINABILITY
Specifications
Includes PTFE cones
Omnifit is a registered trademark of Diba Industries