Learn More
MilliporeSigma™ Supelco™ Ombifit™ Standard Bore Connector Three-way valve connector
Catalog No. 111010332 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Includes O-rings and PTFE cone
Catalog No.
11-101-0332 Includes O-rings and PTFE cone
1 options
Catalog No. 11-101-0332 Supplier MilliporeSigma™ Supelco™ Supplier No. 56034
Add to Cart
Add to Cart
Omnifit™ Standard Bore Connector is very versatile in nature and is used for connecting various types of hardwall tubings. The standard bore connector has a manual valve operation on each end, thus the flow can be switched on or off as required. It is primarily used for fluid-line interconnection, sampling and mixing.
TRUSTED_SUSTAINABILITY
Specifications
Includes O-rings and PTFE cone
Omnifit is a registered trademark of Diba Industries