Learn More
Cytiva (Formerly Pall Lab) Omega Membrane Discs - 50K - 43 mm (12/pkg)
Supplier:  Cytiva OM050043
Encompass
High Performance and Stability for Users of Conventional, Non-Disposable Stirred Cell-Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose. This results in lower processing times and the highest possible recoveries.-Discs can be washed and reused. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3482