Learn More
AdipoGen Omentin (human) (rec.) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B00423010
Encompass_Preferred
Protein. Human omentin (aa 17-313) is fused at the N-terminus to a FLAG-tag. Source: CHO cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). Omentin (Intelectin-1) is highly expressed in omental adipose tissue, small intestine and various other tissues. Omentin has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. It increases AKT phosphorylation in the absence and presence of insulin. It was suggested to play a role in the defense system against microorganisms by recognizing carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues.
Catalog No. 50-205-3117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.