Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma OMI™ Purifier Tube

Catalog No. 111016417 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Hall, ECD, GC/MS Detection Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-101-6417 Hall, ECD, GC/MS Detection Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6417 Supplier MilliporeSigma Supplier No. 23906
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purifier tubes simultaneously remove oxygen, water vapor, CO, CO2, most sulfur compounds, most halogen compounds, alcohols, phenols to less than 10 ppb.

  • OMI™ purifier tubes simultaneously remove oxygen, water vapor, CO, CO2, most sulfur compounds, most halogen compounds, alcohols, phenols to less than 10 ppb.
  • Purify helium, hydrogen, nitrogen, argon and methane.
  • Color change indicates purifier exhaustion.
  • Glass body does not diffuse air or off-gas.
  • Ideal for Hall, ECD and GC/MS detection systems.
  • The OMI is a polishing purifier that removes many contaminants that other upstream purifiers miss.
  • It is recommended that an OMI be installed in carrier gas streams just upstream of every GC.
TRUSTED_SUSTAINABILITY

Specifications

Type Purifier Tube
For Use With (Equipment) Hall, ECD, GC/MS Detection Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.