Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva VIA Extractor™ tissue disaggregator - Omics Bundle

Catalog No. 09927163 Shop All Cytiva Products
Change view
Click to view available options
Voltage:
24 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
09-927-163 24 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-927-163 Supplier Cytiva Supplier No. 29517120
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Semi-automated system for consistent, rapid disaggregation of solid tissue, including tumors, into viable single cells. The VIA Extractor includes tissue disaggregator, controlled-rate freezer and the Omics clamp.

  • The VIA Extractor™ tissue disaggregator disaggregates solid tissue into viable single cells
  • For use in high-throughput omics and flow cytometry research
  • The closed system uses mild processing to optimize yield and preserve content relative to the parent sample
TRUSTED_SUSTAINABILITY

Specifications

Product Type Tissue Disaggregator
Speed ≤460 rpm
Voltage 24 V

For research use only (RUO). Not for diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.