Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diba Omnifit™ Column Frit Removal Tool; 1/ea

Catalog No. OF006090 Shop All Oakton Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
OF006090 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OF006090 Supplier Diba Supplier No. 2194022
Add to Cart
Add to Cart

Remove frits from Omnifit™ chromatography columns

Remove frits from Omnifit™ chromatography columns

Specifications

Product Type Frit Removal Tool
For Use With (Equipment) Chromatography column
Length (Metric) 27.9 cm
Material Acetal Plastic, Stainless Steel
Color Gray
For Use With (Application) Remove frits from Omnifit chromatography columns
No. per Pack 1 Ea.
Sterility Non-sterile
Weight (Metric) 0.09 kg
Width (Metric) 19.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.