Learn More
Diba Omnifit™ Packing Sleeves for EZ and EZ SolventPlus™ Columns
Specifications
Specifications
| Type | Liquid Chromatography Column |
| Diameter (Metric) Inner | 35 mm |
| For Use With (Equipment) | Omnifit™ EZ and EZ SolventPlus™ Columns |
| Length (Metric) | 4 cm |