Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diba Omnifit™ T Series Bottle Vapor Trap and Indicator

Catalog No. 14500719 Shop All Oakton Products
Change view
Click to view available options
No. per Case:
5/Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
14-500-719 5/Pk.
14-500-718 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-500-719 Supplier Diba Supplier No. 2194331
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easy to use

Not recommended for use under pressure with Luer products fitting as no Luer lock is employed.

Order Info

Available with Luer port or Luer port with fluorocarbon check valve and 10μm porosity PTFE inlet filter enabling air to flow into the bottle while preventing particulate contamination and preventing harmful vapors from escaping.

Specifications

Color Blue, White
For Use With (Application) For distilled water, foodstuff
Material PTFE
No. per Case 5/Pk.
Sterility Non-sterile
Neck Finish 1/4-28
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Solvent Waste Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.