Learn More
Medchemexpress LLC HY-135147 5mg Medchemexpress, ONC206 CAS:1638178-87-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1351475MG
Encompass_Preferred
Medchemexpress, HY-135147 5mg ONC206 CAS:1638178-87-6 ONC206 is an analogue of TRAIL inducer ONC201. ONC206 is a selective antagonist of the D2-like dopamine receptors (DRD2/3/4) at nanomolar concentrations. ONC206 has broad-spectrum anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9864
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.