Learn More
ACROBiosystems Onchocerca volvulus OV16 protein, GST Tag
Supplier:  ACROBiosystems OV6V51G4100UG
Encompass_Preferred
Recombinant Protein;100UG;Onchocerca volvulus OV16, GST Tag (OV6-V51G4) is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 (Accession # P31729-1).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5020
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.