Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncrasin 1 95.0+%, TCI America™
SDP

Catalog No. O045010MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O045010MG 10 mg
O045050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O045010MG Supplier TCI America Supplier No. O045010MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75629-57-1
Molecular Formula C16H12ClNO
Molecular Weight (g/mol) 269.73
MDL Number MFCD01051808
InChI Key ZDRQMXCSSAPUMM-UHFFFAOYSA-N
PubChem CID 872445
IUPAC Name 1-[(4-chlorophenyl)methyl]-1H-indole-3-carbaldehyde
SMILES ClC1=CC=C(CN2C=C(C=O)C3=CC=CC=C23)C=C1

Specifications

Melting Point 117°C
Quantity 10 mg
Synonym oncrasin-1, 1-4-chlorobenzyl-1h-indole-3-carbaldehyde, oncrasin 1, 1-4-chlorophenyl methyl indole-3-carbaldehyde, 1-4-chlorophenyl methyl-1h-indole-3-carboxaldehyde, 1-4-chloro-benzyl-1h-indole-3-carbaldehyde, oncrasin1, cambridge id 5601604, n-4-chlorobenzyl-3-formylindole, oncrasin hplc
Formula Weight 269.73
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Oncrasin 1
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.