Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncrasin 1, Tocris Bioscience™

Catalog No. 342710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-271-0 10 mg
34-275-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-271-0 Supplier R&D Systems Supplier No. 3427/10
This item is not returnable. View return policy

Induces abnormal nuclear aggregation of PKCι; proapoptotic

Proapoptotic agent that induces abnormal nuclear aggregation of PKCι and suppresses RNA transcription. Exhibits antiproliferative effects against various human tumor cell lines with K-Ras mutations (IC50 ≤ 3 μM) with minimal effects on normal epithelial cells and inhibits human xenograft growth by 75% in vivo.

Chemical Identifiers

CAS 75629-57-1
Molecular Formula C16H12ClNO
Molecular Weight (g/mol) 269.73
MDL Number MFCD01051808
InChI Key ZDRQMXCSSAPUMM-UHFFFAOYSA-N
Synonym oncrasin-1, 1-4-chlorobenzyl-1h-indole-3-carbaldehyde, oncrasin 1, 1-4-chlorophenyl methyl indole-3-carbaldehyde, 1-4-chlorophenyl methyl-1h-indole-3-carboxaldehyde, 1-4-chloro-benzyl-1h-indole-3-carbaldehyde, oncrasin1, cambridge id 5601604, n-4-chlorobenzyl-3-formylindole, oncrasin hplc
PubChem CID 872445
IUPAC Name 1-[(4-chlorophenyl)methyl]-1H-indole-3-carbaldehyde
SMILES ClC1=CC=C(CN2C=C(C=O)C3=CC=CC=C23)C=C1

Specifications

Quantity 10 mg
Formula Weight 269.73
Percent Purity >98%
Chemical Name or Material Oncrasin 1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.