Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ONE Broth-Salmonella Base

Catalog No. p-6980032
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM1091B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CM1091B Supplier Thermo Scientific™ Supplier No. CM1091B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enrich and recover Salmonella spp. in a single step with Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Base.

Enrich and recover Salmonella spp. from food samples in a single step with Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Base, when prepared with Thermo Scientific Oxoid ONE Broth-Salmonella Selective Supplement (SR0242).

ONE Broth-Salmonella Base is a highly nutritious and selective medium that allows recovery of Salmonella species within 16 to 20 hours. ONE Broth-Salmonella Base requires only a single enrichment medium step. When followed by inoculation onto Thermo Scientific™ Brilliance™ Salmonella Agar and the Thermo Scientific Oxoid Salmonella Latex Test, it forms the Oxoid™ Salmonella Precis™ method which reduces time to result over conventional culture methods. The Oxoid Salmonella Precis method is AFNOR validated.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description ONE Broth-Salmonella Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Species Salmonella
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.