Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ One Shot™ LB Agar Plates with or without Antibiotics

Catalog No. A55803
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 10 Plates
Additive:
50 μg/mL Kanamycin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Additive
A55803 2 x 10 Plates 50 μg/mL Kanamycin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A55803 Supplier Invitrogen™ Supplier No. A55803

One Shot LB Agar Plates are pre-poured plates containing Luria-Bertani (LB) agar medium that are available with or without antibiotics. LB agar is a solid bacterial growth medium used in molecular biology studies for the cultivation and maintenance of Escherichia coli strains. Sterile glass beads are included to aid in plating.

One Shot LB Agar Plates are pre-poured plates containing Luria-Bertani (LB) agar medium that are available with or without antibiotics. LB agar is a solid bacterial growth medium used in molecular biology studies for the cultivation and maintenance of Escherichia coli strains. Autoclaved glass beads are included to aid in plating.

LB agar is a popular, nutrient-rich medium made from casein peptone, yeast extract, sodium chloride, and agar. Casein peptone or tryptone is an enzymatic hydrolysate of casein. It supplies nitrogen, carbon, and amino acids. Yeast extract provides trace elements, vitamins (including B group vitamins), amino acids, and peptides. Sodium chloride supplies sodium cations for membrane transport and maintains osmotic balance. Agar is a solidifying agent of the growth medium.

One Shot LB Agar Plate composition is based on the Jeffrey H. Miller formulation and contains twice as much sodium chloride as the Lennox formulation. The inclusion of autoclaved glass beads allows for simple plate inoculation without the need for additional loops or spreaders.

As E. coli is unable to digest agar but can absorb nutrition from the LB medium, single colonies can be easily isolated and selected. On plates with a selective antibiotic only transformed E. coli with resistance can grow.

One Shot LB Agar Plate formulation

10 g/L casein peptone, 5 g/L yeast extract, 10 g/L NaCl, 15 g/L agar

Benefits of One Shot LB Agar Plates

  • Read-to-use format conveniently packaged in two sleeves of 10 plates
  • Long-shelf life: 3.5 months without antibiotic and 3 months with antibiotic
  • Autoclaved glass plating beads provided for easy cell spreading

One Shot LB Agar Plates are a good choice for use with our Gateway, TOPO, and Gibson Assembly cloning kits and One Shot competent cells to provide faster, reproducible results to support your downstream applications.

Primary applications

  • Selecting transformed coli colonies using antibiotics
  • Streaking and isolating bacteria cells on LB agar plate
  • Bacteriophage plaque assay using a lawn of bacteria

Discover innovative products for your cloning workflow

We offer a wide selection of ready-to-use bacterial growth media formulations, such as LB (Lennox), Terrific Broth, and S.O.C recovery medium. With our selection guide you can explore our One Shot competent cell strains and discover a wide range of cloning solutions for routine and specialty applications, along with our gene synthesis services.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • LB Agar plates with 50 μg/mL Kanamycin, 20 plates
• Autoclaved glass plating beads

Store at 2°C to 14°C.
Additive 50 μg/mL Kanamycin
Format Plate
Manufacturing Origin Spain
Media Type LB Agar (Miller)
Product Type Agar Plate
Sterility Sterile
Target Organism Class E. coli, Coliforms
Fill Volume 21 ± 2 mL
For Use With (Application) Cloning, streaking, and isolating bacteria cells, Bacteriophage plaque assay
Quantity 2 x 10 Plates
Diameter (Metric) 90 mm
Species Escherichia coli
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.