Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ One-Sided Polyester Tags: TARGET ORGANS AFFECTED HEALTH HAZARD: FIRE HAZARD: REACTIVITY: SPECIAL PROTECTION CONSULT CORRESPONDING MSDS FOR FURTHER HAZARDOUS INFORMATION AND INSTRUCTIONS

Catalog No. 19113101 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
25/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-113-101 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-113-101 Supplier Brady™ Supplier No. 99108
Add to Cart
Add to Cart

Identify facility equipment to your exact specifications. Brady™ One-Sided Polyester Tags are durable one-sided tags with a write-on area that accepts most marking pens.

  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant

Specifications

Quantity 25/Pk.
Certifications/Compliance OSHA 1910.145
Color Black, Blue, Red, Yellow, White
Material Polyester
Message TARGET ORGANS AFFECTED HEALTH HAZARD: FIRE HAZARD: REACTIVITY: SPECIAL PROTECTION CONSULT CORRESPONDING MSDS FOR FURTHER HAZARDOUS INFORMATION AND INSTRUCTIONS
Includes (25) nylon fasteners
Shape Rectangle
Temperature Range (English) -40°F to 212°F
Dimensions (L x W) 3 x 5.75 in.
Label Type Not laminated
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.