Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ OneStep TUNEL Apoptosis Kit, FITC
SDP

Catalog No. NB170653 Shop All Novus Products
Change view
Click to view available options
20 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NB170653 20 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170653 Supplier R&D Systems™ Supplier No. NBP31195720ASSAYS

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

Specifications

Description OneStep TUNEL
For Use With (Application) Flow Cytometry, Fluorescence Imaging
Product Type Apoptosis Kit
Content And Storage Store at -20C.
Product Line Novus
Quantity 20 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.