Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ OneStep TUNEL Apoptosis Kit, Red, 594
SDP

Catalog No. NB170656 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170656 100 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170656 Supplier Novus Biologicals™ Supplier No. NBP311959100ASSAYS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Suitable for in situ apoptosis detection of tissue samples (Paraffin embedding, freezing and ultrathin section) and cells (Cell smears, cell climbing smears).

Specifications

Description OneStep TUNEL
Content And Storage Store at -20C.
Quantity 100 Assays
For Use With (Application) Flow Cytometry, Fluorescence Imaging
Product Type Apoptosis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.