Learn More
Dunlop™ Onguard™ Chest Waders with Cleated Outsole
SDP
Superior waterproof protection that resists oil, gas, grease, and saltwater in every condition.
Supplier:  Dunlop™ 860671300
 View more versions of this product
Catalog No. 19-055036
Add to Cart
Description
Description
These durable chest waders are made of heavy duty PVC and boast cleated outsoles for excellent flexibility and traction clogging prevention, even in wet or muddy terrain.
  • Chest high, with PVC upper and non-absorbent polyester backing
  • Injection molded, seamless boot construction
  • Steel midsole and adjustable polyester and nylon strap
  • Excellent cold weather flexibility
  • Can be folded or rolled without cracking
  • Top drawstring closure for snug fit
  • Non-corrosive, reinforced suspender buttons
  • Built-in belt loops with adjustable belt
  • Replaceable cushion insoles
  • Made in the USA
Specifications
Specifications
ASTM F2413-11 M I/75 C/75 SD
Black
PVC, Polyester
13
Polyester/Nylon
Drawstring
Utilities, Municipalities, Aquaculture, and Commercial Fishing
Seamless
Cleated Outsole, Steel Midsole, Replaceable Cushion Insole
Steel
Product Suggestions
Product Suggestions