Learn More
Dunlop™ Onguard™ DuraPro™ Chemical Resistant Steel Toe Boots with Steel Shank
High performance steel toe boot provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole
Supplier:  Dunlop™ 890866
 View more versions of this product
Catalog No. 18-500-517
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
  • 360-degree pull tab for easy donning
  • Larger top circumference for quick and easy pull-on
  • 3/4 in. height inner liner allows  increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
  • Honeycomb design molded inside the heel to reduce impact when walking
  • Polyblend Upper; Sureflex Outsole
  • Ergonomically designed foot-bed to provide better underfoot support
  • Protective bumpers at rear of boot to prevent premature wear/puncture
  • Extended circumference kick-off ledge
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blue
Steel shank
Next Generation Safety-loc
Chemicals
Polyblend, Sureflex
6
Steel toe
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.