Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Pullover Slicker Boots

Catalog No. 19024523 Shop All Onguard Industries LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
8
10
11
12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-024523 11
19-024524 12
19-024522 10
19-024527 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-024523 Supplier Dunlop™ Supplier No. 880501100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fit easily over work shoes

  • Waterproof with excellent cold weather flexibility and chemical resistance
  • Nonconductive adjustable polyester strap with plastic buckle and snaps
  • Nonabsorbent polyester lining
  • 17 in. high (43cm)
  • Yellow with black cleated soles and kick-off lugs

TRUSTED_SUSTAINABILITY

Specifications

Buckle Material Plastic
Color Yellow
For Use With (Application) Construction, concrete work, utilities and food processing
Height (English) 17 in.
Liner Material Polyester
Material Flexothane, PVC
Size 11
Sole Features Black self-cleaning cleated outsole
Strap Material Polyester
Waterproof Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.