Learn More
Dunlop™ Onguard™ Wader Work Boots
Flexible in cold weather
Supplier: Dunlop™ 860560700
Description
- Waterproof hip waders with built-in steel shanks
- Durable PVC uppers with polyester backing
- Can be folded or rolled with no cracking
- Resist oil, grease, gasoline, and salt water
- Nonconductive adjustable polyester-nylon strap with plastic snaps
- Cleated soles
- Black
Compliance
Steel toe meets or exceeds ASTM™ F2413-11
Specifications
ASTM F2413-11 | |
For utilities, municipalities, aquaculture and commercial fishing | |
PVC, Polyester | |
Steel | |
Cleated outsole, Steel midsole, replaceable cushion insole | |
Steel |
Black | |
Plastic snaps, non-absorbent lining | |
Seamless | |
7 | |
Polyester Nylon | |
Oil, grease, gasoline, salt water |