Learn More
Dunlop™ Onguard™ Wader Work Boots
Flexible in cold weather
Supplier:  Dunlop™ 860560900
 View more versions of this product
Catalog No. 19-024386
Only null left
Add to Cart
Description
Description
  • Waterproof hip waders with built-in steel shanks
  • Durable PVC uppers with polyester backing
  • Can be folded or rolled with no cracking
  • Resist oil, grease, gasoline, and salt water
  • Nonconductive adjustable polyester-nylon strap with plastic snaps
  • Cleated soles
  • Black
Compliance
Steel toe meets or exceeds ASTM™ F2413-11
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASTM F2413-11
For utilities, municipalities, aquaculture and commercial fishing
PVC, Polyester
Steel
Cleated outsole, Steel midsole, replaceable cushion insole
Steel
Black
Plastic snaps, non-absorbent lining
Seamless
9
Polyester Nylon
Oil, grease, gasoline, salt water
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.