Learn More
Pyramex™ Onix Plus™ Safety Eyewear
Catalog No. 19155743 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
IR/UV 3.0
IR/UV 5.0
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-743 IR/UV 5.0
19-155-744 IR/UV 3.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-155-743 Supplier Pyramex™ Supplier No. SB4950STP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight frame and suspended lens design provide perfect fit for a variety of users
  • Adjustable nosepiece provides 3 different angles
  • 3-angle adjustable flip lens
  • Unique outer flip lens system provides users with multiple options
  • Flip the outer lens down for applications where tinted or IR lenses are needed
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint IR/UV 5.0
    Frame Color Black
    Weight (Metric) 27 g
    Lens Material Polycarbonate