Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Onix™ Safety Eyewear

Lightweight frame and suspended lens design provide perfect fit for a variety of users. Cushioned temple tips provide non-slip comfort.

$218.78

Specifications

Certifications/Compliance ANSI Z87.1
Weight (Metric) 27 g
Lens Material Polycarbonate
View More Specs

Products 5
Catalog Number Mfr. No. Frame Color Lens Tint Lens Coating Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Lens Tint Lens Coating Price Quantity & Availability  
19-155-957
View Documents
Pyramex™
SGR4910ST
Black, Green Clear Impact-resistant
Case of 12 for $218.78
Only null left
Add to Cart
 
19-155-739
View Documents
Pyramex™
SB4910S
Black Clear N/A N/A N/A
19-155-740
View Documents
Pyramex™
SB4910ST
Black Clear Impact-resistant N/A N/A N/A
19-155-741
View Documents
Pyramex™
SB4920S
Black Gray N/A N/A N/A
19-155-974
View Documents
Pyramex™
SP4910S
Black, Pink Clear N/A N/A N/A
Description

Description

  • Suspended lens provides stylish look, comfort fit, panoramic view and maximum protection
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Adjustable nose piece provides 3 different angles, allowing the user to find the most comfortable fit
  • Ratcheting temples provide different angles for perfect view and fit
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications

    Specifications

    ANSI Z87.1
    27 g
    Polycarbonate
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.