Learn More
Selleck Chemical LLC Tamnorzatinib S8933-5MG
SDP
Supplier:  Selleck Chemical LLC S89335MG
Encompass_Preferred
Tamnorzatinib (ONO-7475) is a potent selective and orally active novel inhibitor of Anexelekto(Axl)/MER tyrosine kinase with IC50 of 0 7 nM and 1 0 nM for AXL and MER respectively ONO-7475 suppresses the emergence and maintenance of tolerant cells to the initial EGFR-TKIs osimertinib or dacomitinib in AXL-overexpressing EGFR-mutated NSCLC cells ONO-7475 arrests growth and kills FMS-like tyrosine kinase 3-internal tandem duplication mutant acute myeloid leukemia cells
Catalog No. 50-217-3443
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.