Learn More
Apexbio Technology LLC ONO-AE3-208 402473-54-5 25mg
SDP
Supplier:  Apexbio Technology LLC A368425
Encompass_Preferred
ONO-AE3-208 (CAS 402473-54-5) is a selective antagonist of the prostaglandin E2 receptor subtype EP4 a receptor implicated in inflammatory diseases and cancer progression particularly prostate cancer In cellular studies ONO-AE3-208 inhibits migration and invasion of prostate cancer cell lines (PC3 and LNCaP) reducing wound healing rates at 10 M and suppressing invasion in Transwell assays at 0 1 M In vivo the compound has been reported to limit bone metastasis of prostate cancer cells Additionally ONO-AE3-208 has been used in models of abdominal aortic aneurysm where it exhibits dose-dependent inhibition of elastin degradation and aneurysm formation
Catalog No. 50-000-96765
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.