Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ononetin
GREENER_CHOICESDP

Catalog No. 514350
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-435-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-435-0 Supplier Tocris Bioscience™ Supplier No. 5143/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TRPM3 blocker

Naturally occurring deoxybenzoin. TRPM3 channel blocker (IC50 = 300 nM). Inhibits pregnenolone sulfate-induced intracellular Ca2+ accumulation in HEKmTRPM3 cells and dorsal root ganglia (DRG) neurons in vitro.

Chemical Identifiers

CAS 487-49-0
Molecular Formula C15H14O4

Specifications

Quantity 50 mg
Formula Weight 258.27
Molecular Weight (g/mol) 258.273
Chemical Name or Material Ononetin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.