Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Warner Instruments™ Open Bath Recording and Imaging Chambers

Catalog No. BTX640223 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
Bath Volume:
70 μL
133 μL
150 μL
180 μL
234 μL
560 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bath Volume
BTX640223 70 μL
BTX640235 234 μL
BTX640241 560 μL
BTX640233 133 μL
BTX640236 180 μL
BTX640234 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. BTX640223 Supplier Warner Instruments™ Supplier No. 640223
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for physiological experiments performed with inverted or dissecting microscopes, including patch and oocyte recording, isolated perfused tubule, and cell impalement studies. RC chambers have also been used with fluorescence microscopy.

  • Polycarbonate RC Series recording chambers
  • Uses a user-attached #2 coverslip, 24 x 60 mm, as the bath floor
  • Coverslip attaches with vacuum grease, Sylgard, or RTV cement
  • Models vary by bath diameter, shape, and profile height
  • Supports gravity-fed or pump-controlled solution input and output
  • Gravity-feed flow rates can exceed 10 mL/min
  • Perfusion input accepts 1.57 mm OD polyethylene tubing - PE-160
  • Separate suction reservoir reduces recording-chamber noise
  • Adjustable suction tube controls the bath solution level
  • Compatible with MP Series manifolds for multiple perfusion lines, including part number 64-0211
  • Includes a single-line connector
  • Isolated reservoir supports an agar bridge in the recording ground circuit

Specifications

Compatibility 15 mm Coverglass
For Use With (Equipment) Imaging Chamber
Bath Volume 70 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.