Learn More
Description
- Polycarbonate RC Series recording chambers
- Uses a user-attached #2 coverslip, 24 x 60 mm, as the bath floor
- Coverslip attaches with vacuum grease, Sylgard, or RTV cement
- Models vary by bath diameter, shape, and profile height
- Supports gravity-fed or pump-controlled solution input and output
- Gravity-feed flow rates can exceed 10 mL/min
- Perfusion input accepts 1.57 mm OD polyethylene tubing - PE-160
- Separate suction reservoir reduces recording-chamber noise
- Adjustable suction tube controls the bath solution level
- Compatible with MP Series manifolds for multiple perfusion lines, including part number 64-0211
- Includes a single-line connector
- Isolated reservoir supports an agar bridge in the recording ground circuit
Specifications
Specifications
| Compatibility | 24 x 50 mm Coverglass |
| For Use With (Equipment) | Imaging Chamber |
| Bath Volume | 560 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.