Learn More
Ampco Safety Tools Open End Wrench (Imperial), < 2 in.
AMPCO™ Open End Wrench performs in high-risk and demanding environments.
$98.00 - $1124.00
Specifications
Head Style | Open End |
---|---|
Material | Aluminum Bronze |
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog Number | Mfr. No. | Length (English) Opening | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) Opening | Price | Quantity & Availability | |||||
19-173-228
![]() |
Ampco Safety Tools
0250 |
5/16 in. |
Each for $112.40
|
|
|||||
19-173-229
![]() |
Ampco Safety Tools
0252 |
11/32 in. |
Each for $109.20
|
|
|||||
19-173-230
![]() |
Ampco Safety Tools
0254 |
3/8 in. |
Each for $98.00
|
|
|||||
19-173-231
![]() |
Ampco Safety Tools
0256 |
13/32 in. |
Each for $127.20
|
|
|||||
19-173-232
![]() |
Ampco Safety Tools
0258 |
7/16 in. |
Each for $134.70
|
|
|||||
19-173-233
![]() |
Ampco Safety Tools
0260 |
1/2 in. |
Each for $162.20
|
|
|||||
19-173-234
![]() |
Ampco Safety Tools
0262 |
9/16 in. |
Each for $182.40
|
|
|||||
19-173-235
![]() |
Ampco Safety Tools
0264 |
19/32 in. |
Each for $204.00
|
|
|||||
19-173-236
![]() |
Ampco Safety Tools
0266 |
5/8 in. |
Each for $191.90
|
|
|||||
19-173-237
![]() |
Ampco Safety Tools
0268 |
11/16 in. |
Each for $231.50
|
|
|||||
19-173-238
![]() |
Ampco Safety Tools
0270 |
3/4 in. |
Each for $246.00
|
|
|||||
19-173-239
![]() |
Ampco Safety Tools
0272 |
25/32 in. |
Each for $272.50
|
|
|||||
19-173-240
![]() |
Ampco Safety Tools
0274 |
13/16 in. |
Each for $312.00
|
|
|||||
19-173-241
![]() |
Ampco Safety Tools
0276 |
7/8 in. |
Each for $312.00
|
|
|||||
19-173-242
![]() |
Ampco Safety Tools
0278 |
15/16 in. |
Each for $350.00
|
|
|||||
19-173-243
![]() |
Ampco Safety Tools
0280 |
31/32 in. |
Each for $350.00
|
|
|||||
19-173-244
![]() |
Ampco Safety Tools
0282 |
1 in. |
Each for $403.00
|
|
|||||
19-173-245
![]() |
Ampco Safety Tools
0284 |
1-1/16 in. |
Each for $403.00
|
|
|||||
19-173-246
![]() |
Ampco Safety Tools
0286 |
1-1/8 in. |
Each for $453.00
|
|
|||||
19-173-247
![]() |
Ampco Safety Tools
0288 |
1-3/16 in. |
Each for $453.00
|
|
|||||
19-173-248
![]() |
Ampco Safety Tools
0290 |
1-1/4 in. |
Each for $524.00
|
|
|||||
19-173-249
![]() |
Ampco Safety Tools
0292 |
1-5/16 in. |
Each for $524.00
|
|
|||||
19-173-250
![]() |
Ampco Safety Tools
0294 |
1-3/8 in. |
Each for $615.00
|
|
|||||
19-173-251
![]() |
Ampco Safety Tools
0296 |
1-7/16 in. |
Each for $615.00
|
|
|||||
19-173-252
![]() |
Ampco Safety Tools
0298 |
1-1/2 in. |
Each for $615.00
|
|
|||||
19-173-253
![]() |
Ampco Safety Tools
0300 |
1-9/16 in. |
Each for $740.00
|
|
|||||
19-173-254
![]() |
Ampco Safety Tools
0302 |
1-5/8 in. |
Each for $740.00
|
|
|||||
19-173-255
![]() |
Ampco Safety Tools
0304 |
1-11/16 in. |
Each for $939.50
|
|
|||||
19-173-256
![]() |
Ampco Safety Tools
0306 |
1-3/4 in. |
Each for $939.50
|
|
|||||
19-173-257
![]() |
Ampco Safety Tools
0308 |
1-13/16 in. |
Each for $939.50
|
|
|||||
19-173-258
![]() |
Ampco Safety Tools
0310 |
1-7/8 in. |
Each for $1,124.00
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Open End | |
Aluminum Bronze | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.