Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Open-Finger Shock Absorbing Glove Liners

Catalog No. 189994599B Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Hand:
Right
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
18-999-4599B Medium Right
18-999-4599A Small Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-4599B Supplier Impacto™ Supplier No. 50701310032
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VEP padding throughout the glove liner offers protection from shock and impact. Use under work gloves in confined spaces to protect the back of the hand.

  • Breathable Nylon Lycra™ is designed to be worn inside work gloves
  • Padded with 3/16 in. impact and shock absorbing VEP pad
  • Open finger design provides tactile feedback
  • Ideal for use with hand tools, or in confined space applications such as mining

Specifications

Color Black
Hand Right
Material Nylon Lycra
Product Type Glove Liners
Size Medium
Finger Style Three Quarter
For Use With (Application) Manufacturing Tools, and Confined Spaces
Glove Cuff Style Open
Series 507-01
Length (English) 9 in.
Length (Metric) 22.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.