Learn More
MilliporeSigma™ Supelco™ Open-Top Screw Cap
Catalog No. 111012011 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Closure Size:
15-425
24-400
8-425
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
11-101-2011 24-400
11-101-2081 15-425
11-101-2113 8-425
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-2011 Supplier MilliporeSigma™ Supelco™ Supplier No. 27057
Add to Cart
Add to Cart
These hole caps do not come with septa.
TRUSTED_SUSTAINABILITY
Specifications
Description Black polypropylene hole cap
Material Polypropylene
No. per Pack 100
Closure Size 24-400
For Use With (Equipment) 20, 40 or 60 mL Vial