Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Open Your Own Geode
SDPSCI_ED

Catalog No. S79092
Encompass
Encompass_Preferred
Change view
Click to view available options
Specimen Size:
2.54 x 3.81 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Specimen Size
S79092 2.54 x 3.81 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S79092 Supplier Geoscience Industries Supplier No. M100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Two unopened geodes and instructions

Open geodes in the classroom.

  • Amaze students when you open geodes and see the magnificent crystals inside
  • Specimen size: 1 to 1.5 in. (2.54 to 3.81cm)

Specifications

Includes Two unopened geodes and instructions
Specimen Size 2.54 x 3.81 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.