Learn More
AdipoGen Opipramol
SDP
Supplier:  AdipoGen CDXO0042M050
Encompass_Preferred
Chemical. CAS 315-72-0. Formula C23H29N3O. MW 363.5. Synthetic. Tricyclic antidepressant and anxiolytic compound. Acts as a high affinity sigma receptor agonist, primarily at the sigma1 subtype, but also at the sigma2 subtype with somewhat lower affinity. Shows a low to moderate antagonistic affinity for the D2, 5-HT2, H1, H2, and muscarinic acetylcholine receptors. H1 and H2 receptor antagonism account for its antihistamine effects, and muscarinic acetylcholine receptor antagonism is responsible for its anticholinergic properties.
Catalog No. 50-256-0332
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.