Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPRM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
SDP

Catalog No. p-7223786 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8575 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8575 Supplier Proteintech Group Inc Supplier No. 276251AP150UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The OPRM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OPRM1. This antibody recognizes human, mouse, rat antigen. The OPRM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen OPRM1
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 267 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 4988
Target Species Human, Rat, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.