Learn More
Apexbio Technology LLC Oprozomib (ONX-0912), 100mg. Cas: 935888-69-0 MFCD: MFCD25976563. Proteasome inhibitor
SDP
Supplier:  Apexbio Technology LLC A1934100
Encompass_Preferred
ONX-0912 (as known as oprozomib), discovered during a medicinal chemistry effort using tripeptide epoxyketones, is an orally bio-available and potent proteasome inhibitor that inhibits growth and induces apoptosis in bortezomib resistant multiple myeloma (MM) cells. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4727
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.