Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Opsin Red/Green Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674400 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-400 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-400 Supplier Invitrogen™ Supplier No. PA19517
Add to Cart
Edge
Add to Cart

Chicken Polyclonal Antibody

PA1-9517 detects Opsin in mouse, rat, zebra finch and tiger samples. PA1-9517 has been successfully used in Western blot procedures. By Western blot, PA1-9517 detects a 44 kDa protein representing Opsin. PA1-9517 immunogen is a synthetic peptide based on the mouse opsin1 protein.

Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal. Abs(max)=530 nm; The three color pigments are found in the cone photoreceptor cells. Phosphorylated on some or all of the serine and threonine residues present in the C-terminal region. Defects in OPN1MW are the cause of partial colorblindness deutan series (CBD) also called deuteranopia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opsin Red/Green
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene OPN1MW
Gene Accession No. O35476, O35599
Gene Alias AW551857; Bcp; blue cone opsin; blue cone opsin-like pigment; blue cone photoreceptor pigment; blue cone pigment; blue opsin; blue/UV opsin; blue-sensitive opsin; BOP; CBBM; CBD; CBP; COD5; cone dystrophy 5 (X-linked); Gcp; GOP; Green cone photoreceptor pigment; green cone pigment; green long wavelength sensitive cone opsin; green LWS photopigment; green-sensitive opsin; Long-wave-sensitive opsin 1; M opsin; Medium wavelength-sensitive cone opsin; medium-wave-sensitive opsin 1; Medium-wave-sensitive opsin 2; Medium-wave-sensitive opsin 3; midwavelength sensitive opsin; MWS opsin; OPN1LW; Opn1mw; OPN1MW1; OPN1MW2; OPN1MW3; Opn1sw; OPSG; opsin 1 (cone pigments), long-wave-sensitive; opsin 1 (cone pigments), medium-wave-sensitive; opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan); opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan), green opsin; opsin 1 (cone pigments), short-wave-sensitive; opsin 1 (cone pigments), short-wave-sensitive (color blindness, tritan); Opsin 1 cone pigments medium-wave-sensitive 2; opsin 1 cone pigments medium-wave-sensitive 3; Opsin red; OPSR; photopigment apoprotein; RCP; red cone opsin; red cone photoreceptor pigment; red opsin; red-sensitive opsin; ROP; RP23-171N20.2; S opsin; short wavelength sensitive opsin; short wavelength-sensitive cone opsin; Short-wave-sensitive opsin 1; SWS opsin; UV cone pigment
Gene Symbols OPN1LW, OPN1MW
Host Species Chicken
Immunogen Synthetic peptide mixture corresponding to residues 14-26 opsin1 of mouse green cone and residues 18-30 of tiger red cone.
Purification Method IgY fraction
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14539, 20164, 89810
Target Species Mouse, Rat, Avian
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.