Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Opti-Clear Eye Drops

Catalog No. 19090642 Shop All Medique Products
Change view
Click to view available options
Quantity:
0.5 oz. Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-642 0.5 oz. Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-642 Supplier Medique Supplier No. 49566

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Provides fast, soothing relief to irritated eyes.

  • Contains tetrahydrozoline

Specifications

For Use With (Application) Fast, Soothing Relief For Irritated Eyes
Product Type Eye Drops
Quantity 0.5 oz. Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.