Learn More
Optimize Technologies OPTI-GUARD 3mm Guard Cartridge, C8, 3pk
SDP
Supplier:  Optimize Technologies 100201485
Encompass_Preferred
The OPTI-GUARD™ 3mm maintains the tool-free connectivity of the 1mm, but incorporates a cartridge based format to allow for a larger bed with more capacity. The two-part holder is designed for use with any analytical column to provide vital protection from dirty samples and strongly retained contaminants. An auto-adjusting stem conforms to any tube stop depth for a perfect zero-dead-volume connection. Connecting the OPTI-GUARD™ requires no extra tubing, maintaining the minimal impact philosophy that has made the OPTI-GUARD™ line so popular. Key Features include: 3 disposable cartridges, automatic adjusting to any manufacturer's column, perfect repeat ZDV connection, minimal impact - no change in efficiency, provides vital protection from dirty samples and strongly retained contaminants. Use with OPTI-GUARD™ 3mm Holder 10-02-01495.
Catalog No. 50-113-230
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.