Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ OPTI-GUARD™ C18 (Violet Label) 1 mm Guard Column

Catalog No. 111009001 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter: 1 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-100-9001 Diameter: 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9001 Supplier MilliporeSigma™ Supelco™ Supplier No. 51177U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OPTI-GUARD™ sets the standard for low-impact, easy to use pre-column protection.

Designed for use with analytical (4.6 mm, 3.0 mm I.D.) and narrow bore (2.1 mm, 1.0 mm) columns, the patented floating stem design automatically adjusts to any manufactures tube stop depth for a zero-dead volume connection every time. The OPTI-GUARD™ 1 mm requires no special connecting hardware or tools for installation.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5/Pk.
Type Guard Column
Phase Reversed Phase
For Use With (Application) HPLC
Product Line OPTI-GUARD

OPTI-GUARD is a registered trademark of Optimize Technologies, Inc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.