Learn More
Optimize Technologies OPTI-SEAL UHMW-PE Piston Seal, Varian 2010, 2510
SDP
Supplier:  Optimize Technologies 383601000
Encompass_Preferred
Optimize Piston Seals are the longest lasting seals on the market. Machined from either of two proprietary polymer blends, UHMW-PE or ITB, Optimize Seals provide the ultimate in high pressure sealing capacity. For mostly aqueous buffered mobile phases, Opti-Seal™ UHMW-polyethylene is the wisest choice. Specially designed to be more resistant to abrasion and wear than standard polyethylene seals, they can last up to seven times longer under mostly aqueous conditions. Despite the UHMW-PE's durability, it has a coefficient of friction approaching that of PTFE. The homogenous polymer nature leads to less shedding, but when it does occur, a larger, more fibrous particle is produced which is easily caught by a filter frit. This item is equivalent to OEM part number 00-997261-37.
Catalog No. 50-114-069
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.