Learn More
Puritan™ Opti- Tranz™ Liquid Stuart Media Transport Tube with Miniature Rayon Tip Swab
Description
Specifications
Specifications
| Product Type | Transport Tube |
| Media Type | Semi-solid (Sodium Thioglycollate and Agar) |
| Sterility | Sterile |
| Swab Configuration | Dual Applicator |
| Closure Color | White |
| Closure Type | Screw Cap |
| Description | Diameter: 5.15mm |
| Diameter (Metric) | 5.15 mm |
| For Use With (Application) | Throat, urogential, and wound swab sampling |
| Handle Material | Polystyrene |
| Show More |
For in vitro diagnostic (IVD) use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.