Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles

Catalog No. 09981006
Change view
Click to view available options
Diameter (Metric):
0.1 μm
0.2 μm
0.3 μm
0.4 μm
0.6 μm
0.85 μm
1.25 μm
2.5 μm
Quantity:
15 mL
100 mL
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity
09-981-006 0.3 μm 15 mL
09-980-853 1.25 μm 15 mL
09-980-854 1.25 μm 100 mL
09-981-015 2.5 μm 15 mL
09-981-016 2.5 μm 100 mL
09-981-000 0.1 μm 15 mL
09-981-001 0.1 μm 100 mL
09-981-003 0.2 μm 15 mL
09-981-004 0.2 μm 100 mL
09-981-007 0.3 μm 100 mL
09-980-855 0.4 μm 15 mL
09-980-856 0.4 μm 100 mL
09-980-841 0.6 μm 15 mL
09-980-842 0.6 μm 100 mL
09-980-857 0.85 μm 15 mL
09-980-858 0.85 μm 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 09-981-006 Supplier Thermo Scientific™ Supplier No. 81000797100290
Add to Cart
Add to Cart

Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles.

Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles for use in turbidimetric/nephelometric assays, slide agglutination assays, chemiluminescent assays and in clinical diagnostics and molecular biology applications. Particles can be used direct from the bottle, without any pre-cleaning.

Our production does not utilize common surfactants (like SDS, Tween™ 20, Triton™ X-100) that can interfere with protein binding to particle surfaces. Polystyrene surfaces are very hydrophobic and adsorb proteins almost instantaneously. Our proprietary anionic surfactant does not interfere with the binding of proteins nor cause proteins to desorb from microparticle surfaces.
  • Available in a wide range of diameters (from 0.1 μM to 2.5 μM)
  • No pre-washing
  • Tight size distribution—CV<2%
  • Optimized colloidal stability
  • Shelf-life of 10 years

Recommended for:

Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays

Note:

When using Opti-Bind particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Includes Certificate of Analysis
Content And Storage 2°C to 8°C
Description Opti-Bind Polystyrene Sulfate Particles
For Use With (Application) Diagnostic Assays
Material Polystyrene Sulfate
Product Type Specialty Particles
Includes Package Insert and Certificate of Analysis
Quantity 15 mL
Sample Evaluation Packs These evaluation packs allow you to buy small samples from among our standard inventory of different surfaces, colors, sizes and binding capacities at greatly reduced prices. Opti-Bind particles are packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available Opti-Bind polystyrene particles.
Specialty Packaging Opti-Bind Polystyrene particles are available in different concentrations and package sizes upon request.
Diameter (Metric) 0.3 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.